Learn More
Trajan Scientific SILTITE MICRO UNION
SDP
Supplier:  Trajan Scientific 073560
Encompass_Preferred
Trajan SilTite GC Capillary Column Connectors: u Unions, Connections: 0.1-0.25 mm and 0.1-0.25 mm, For Use With: GC Capillary Column, Includes: 2 unions, 5 double taper ferrules and tools
Catalog No. 50-253-0197