Learn More
Trajan Scientific SILTITE MICRO UNION
SDP
Catalog No. 502530196
Encompass_Preferred
Trajan SilTite GC Capillary Column Connectors: u Unions, Connections: 0.32 mm and 0.32 mm, For Use With: GC Capillary Column, Includes: 2 unions, 5 double taper ferrules and tools
Catalog No. 50-253-0196 Supplier Trajan Scientific Supplier No. 073563
Trajan SilTite GC Capillary Column Connectors: u Unions, Connections: 0.32 mm and 0.32 mm, For Use With: GC Capillary Column, Includes: 2 unions, 5 double taper ferrules and tools