Learn More
Trajan Scientific SILTITE MICRO UNION FRRULE10PK
SDP
Catalog No. 502530193
Encompass_Preferred
Trajan GC Capillary Column Connectors: Replacement Ferrules, For Use With: 0.32 mm I.D. First Column and Second Column with 0.5 mm Holes, Replacement SilTite u-Union Ferrules, Stainless Steel
Catalog No. 50-253-0193 Supplier Trajan Scientific Supplier No. 073569
Trajan GC Capillary Column Connectors: Replacement Ferrules, For Use With: 0.32 mm I.D. First Column and Second Column with 0.5 mm Holes, Replacement SilTite u-Union Ferrules, Stainless Steel