Learn More
Trajan Scientific SILTITE MICRO UNION FRRULE10PK
SDP
Supplier:  Trajan Scientific 073566
Encompass_Preferred
Trajan GC Capillary Column Connectors: Replacement Ferrules, For Use With: 0.1 to 0.25 mm I.D. First and Second Columns with 0.4 mm Holes, Replacement SilTite u-Union Ferrules, Stainless Steel
Catalog No. 50-253-0195