Learn More
Trajan Scientific SilTite Nut Agilent Medical 5/PK
SDP
Catalog No. 5000493958
Encompass_Preferred
SilTite metal nuts for Agilent split/splitless injectors PK5
Catalog No. 50-004-93958 Supplier Trajan Scientific Supplier No. 073226
SilTite metal nuts for Agilent split/splitless injectors PK5