Learn More
Trajan Scientific SilTite Nut Shimadzu Medical 5/PK
SDP
Catalog No. 5000494243
Encompass_Preferred
SilTite metal nuts - Shimadzu QP5000/5050 standard MS Interface PK5
Catalog No. 50-004-94243 Supplier Trajan Scientific Supplier No. 073233
SilTite metal nuts - Shimadzu QP5000/5050 standard MS Interface PK5