Learn More
Trajan Scientific SilTite Nut Shimadzu Medical 5/PK
SDP
Supplier:  Trajan Scientific 073232
Encompass_Preferred
SilTite metal nuts - slotted female thread M5 for use on Shimadzu GC14A GC17A GC2010 and GC2014 detector/injectors (not for MS interfaces or QP2010 injector) PK5
Catalog No. 50-004-94328