Learn More
Trajan Scientific SilTite Union Micro Medical 1/EA
SDP
Catalog No. 5000494454
Encompass_Preferred
SilTite -union for joining 0 32 mm and 0 53 mm columns/fused silica
Catalog No. 50-004-94454 Supplier Trajan Scientific Supplier No. 073564
SilTite -union for joining 0 32 mm and 0 53 mm columns/fused silica