Learn More
Trajan Scientific SilTite Union Micro Medical 10/PK
SDP
Catalog No. 5000494594
Encompass_Preferred
Replacement SilTite -union ferrules for joining 0 53 mm and 0 53 mm columns/fused silica PK10
Catalog No. 50-004-94594 Supplier Trajan Scientific Supplier No. 073571
Replacement SilTite -union ferrules for joining 0 53 mm and 0 53 mm columns/fused silica PK10