Learn More
Trajan Scientific SilTite Union Micro Medical 10/PK
SDP
Catalog No. 5000493929
Encompass_Preferred
Replacement SilTite -union ferrules for joining 0 32 mm and 0 53 mm columns/fused silica PK10
Catalog No. 50-004-93929 Supplier Trajan Scientific Supplier No. 073570
Add to Cart
Add to Cart
Replacement SilTite -union ferrules for joining 0 32 mm and 0 53 mm columns/fused silica PK10