Learn More
Trajan Scientific SilTite Union Micro Medical 5/PK
SDP
Supplier:  Trajan Scientific 073573
Encompass_Preferred
Replacement SilTite -union only (no ferrules) for joining 0 1 - 0 32 mm and 0 53 mm columns/fused silica PK5
Catalog No. 50-004-94913