Learn More
Trajan Scientific SilTite Union Micro Medical 5/PK
SDP
Catalog No. 5000494595
Encompass_Preferred
Replacement SilTite -union only (no ferrules) for joining 0 53 mm and 0 53 mm columns/fused silica PK5
Catalog No. 50-004-94595 Supplier Trajan Scientific Supplier No. 073574
Replacement SilTite -union only (no ferrules) for joining 0 53 mm and 0 53 mm columns/fused silica PK5