Learn More
Sigma Aldrich Fine Chemicals Biosciences SILu™MAb Adalimumab Stable-Isotope Labeled Monoclonal Antibody
Supplier:  Sigma Aldrich Fine Chemicals Biosciences MSQC11100UG
Encompass
SILu™ MAb Adalimumab Stable-Isotope Labeled Monoclonal Antibody is a recombinant, stable isotope labeled, monoclonal antibody which incorporates [13C6, 15N4]-Arginine and [13C6, 15N2]-Lysine. Expressed in CHO cells, SILuMAb Adalimumab is designed to be used as an internal standard for analysis of Adalimumab in human serum. Each vial of SILuMAb Adalimumab contains the labeled antibody lyophilized from a solution of phosphate buffered saline. Vial content was determined by measuring A280 and using an extinction coefficient (E0.1%) of 1.4.
Catalog No. NC1599733