Learn More
Sigma Aldrich Fine Chemicals Biosciences SILuTMMAb K1 Pharmacokinet
Catalog No. 502907802
Encompass
All critical reagents necessary to perform the assay are provided in the kit. The plate-based format avoids the high cost and potential LC column blockage associated with bead-based enrichment formats. Sample preparation with the SILu(TM)MAb PK kit can be performed in under 5 hours and consists of coating wells with high-specificity capture antibody followed by immunoaffinity enrichment and rapid in-plate trypsin digestion. The resulting digest is ready for injection with no SPE or other cleanup required.
Catalog No. 50-290-7802 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. MSKT00011KT
All critical reagents necessary to perform the assay are provided in the kit. The plate-based format avoids the high cost and potential LC column blockage associated with bead-based enrichment formats. Sample preparation with the SILu(TM)MAb PK kit can be performed in under 5 hours and consists of coating wells with high-specificity capture antibody followed by immunoaffinity enrichment and rapid in-plate trypsin digestion. The resulting digest is ready for injection with no SPE or other cleanup required.