Learn More
Hach Company Silver 1 Reagent Powder Pillows, pk/50
Supplier:  Hach Company 2293566
Encompass
Reagent for determination of Silver by the Cadion 2B Colorimetric Method. Hach Method 8120. Range: 0.02 - 0.70 mg/L Ag. Sample Size: 50 mL. Pack of 50 Pillows. Also available in reagent set 2296600.
Catalog No. 50-199-5745