Learn More
Strem, An Ascensus Company CAS# 26077-31-6. 5g. Silver 2-ethylhexanoate, 99%. MFCD00058700
SDP
Catalog No. 5090115422
Encompass
CAS# 26077-31-6. 5g. Silver 2-ethylhexanoate, 99%. MFCD00058700. Molecular Weight: 251.08. Molecular Formula: AgOOCCH(C2H5)C4H9. Color/form: white to off-white pwdr. Strem# 93-4741. http://www.strem.com/catalog/v/93-4741/
Catalog No. 50-901-15422 Supplier Strem, An Ascensus Company Supplier No. 9347415G
CAS# 26077-31-6. 5g. Silver 2-ethylhexanoate, 99%. MFCD00058700. Molecular Weight: 251.08. Molecular Formula: AgOOCCH(C2H5)C4H9. Color/form: white to off-white pwdr. Strem# 93-4741. http://www.strem.com/catalog/v/93-4741/