Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver AA Standard, 1000 ppm Ag in 3% HNO3, Ricca Chemical
SDP

Catalog No. AAG1KN100 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Amber Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAG1KN-100 100 mL Amber Poly Bottle
AAG1KN-500 500 mL Amber Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAG1KN-100 Supplier Ricca Chemical Company Supplier No. AAG1KN100

Chemical Identifiers

CAS 7761-88-8 , 7697-37-2 , 7761-88-8
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
PubChem CID 24470
ChEBI CHEBI:32130
IUPAC Name silver(1+) nitrate
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

Packaging Amber Poly Bottle
Concentration or Composition Notes 1,000ppm Ag
CAS Min % 94.0, 4.22, 0.15
CAS Max % 97.0, 4.39, 0.15
Equivalence 1 mL = 1 mg Ag
Physical Form Liquid
Color Colorless
pH < 2
Melting Point Approximately 0°C
Quantity 100 mL
Chemical Name or Material Silver AA Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.