Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Silver acetate

Catalog No. 20437410G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-437-410G 10 g
20-437-450G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 20-437-410G Supplier Sigma Aldrich Supplier No. 20437410G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Used in a novel preparation of highly reflective, conductive silvered polymer films. Also effectively catalyzes cycloaddition reactions of isocyanoacetates with a variety of olefins.

Specifications

CAS 563-63-3
Molecular Formula C2H3AgO2
Linear Formula CH3COOAg
Quantity 10 g
MDL Number MFCD00012446
Synonym Acetic acid silver salt
Molecular Weight (g/mol) 166.91
Percent Purity 99.99% trace metals basis
Biological Activity Respiratory system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.