Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver Acetate, Powder, Purified, 99%, Spectrum™ Chemical
SDP

Catalog No. 18613115 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-613-115 25 g Amber Glass Bottle
18-613-116 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-613-115 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S104525GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

Chemical Identifiers

CAS 563-63-3

Specifications

Color White
CAS Max % 1
Quantity 25 g
Linear Formula CH3COOAg
Solubility Information Solubility in Water: 10.2 g/L (20°C).
InChI Key WJCVKPGZQKODGL-UHFFFAOYSA-L
Formula Weight 166.91 g/mol
Percent Purity 99%
Grade Purified
Odor Odorless
Packaging Amber Glass Bottle
Appearance White Powder
Show More Show Less
Recommended Storage Light Sensitive, Store In a dark place

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.