Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver Acetate, purum p.a, ≥99.0% (T), Solstice

Catalog No. 60047693 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-693 50 g Glass bottle
60-047-692 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-047-693 Supplier Solstice Supplier No. 8514050G

missing translation for 'validMainframeMsg'

Purum p.a.,≥99.0% (T)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 563-63-3
Molecular Formula C2H3AgO2
Molecular Weight (g/mol) 166.912
MDL Number MFCD00012446
InChI Key CQLFBEKRDQMJLZ-UHFFFAOYSA-M
Synonym silver acetate, silver i acetate, silver 1+ acetate, silver monoacetate, unii-19pps85f9h, acetic acid, silver 1+ salt, acetic acid silver salt, silver 1+ ion acetate, acetic acid, silver 1+ salt 1:1, silver acetate ag oac
PubChem CID 11246
IUPAC Name silver;acetate
SMILES CC(=O)[O-].[Ag+]

Specifications

Quantity 50 g
Linear Formula CH3COOAg
UN Number UN3077
Beilstein 3595636
Formula Weight 166.91g/mol
Percent Purity ≥99.0% (T)
Packaging Glass bottle
Chemical Name or Material Silver acetate
Hazard Statement P261-P273-P305 + P351 + P338
DOTInformation 9 / PGIII
RTECSNumber 209-254-9
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.