Learn More
Bellco Glass, Inc. Silver Alum Seal PTFE/Sil,11mm
SDP
Catalog No. 501945411
Encompass_Preferred
Larger opening provides increased target area for needle penetration. MADE IN THE USA. Size: 11mm Septa material: ptfe silicone Seal color: silver
Catalog No. 50-194-5411 Supplier Bellco Glass, Inc. Supplier No. 207005000
Larger opening provides increased target area for needle penetration. MADE IN THE USA. Size: 11mm Septa material: ptfe silicone Seal color: silver