Learn More
Bellco Glass, Inc. Silver AlumSeal W/PTFE/RR,11mm With PTFE/ Red Rubber Septa
SDP
Catalog No. 501945398
Encompass_Preferred
Larger opening provides increased target area for needle penetration. Available in a variety of colors for easy sample identification. MADE IN THE USA. Size: 11mm Septa: clear ptfe or red rubber
Catalog No. 50-194-5398 Supplier Bellco Glass, Inc. Supplier No. 207002000
Larger opening provides increased target area for needle penetration. Available in a variety of colors for easy sample identification. MADE IN THE USA. Size: 11mm Septa: clear ptfe or red rubber