Learn More
Electron Microscopy Sciences Silver Bioguard Anti-Microbial Mat, 15" x 10"
SDP
Catalog No. 501902669
Encompass_Preferred
Lab mats with nanoscale silver particles bind with proteins to neutralize 99.9% of common microbes. Ensure consistant sample reproduction and results by significantly minimizing workspace contamination. Silver is used extensively in medical applications where microbial contamination is a concern. Mats are less than 1 mm thick and are backed with an anti-skid coating making them perfect for the benchtop or culture hood.
Size: 15" x 10"
Catalog No. 50-190-2669 Supplier Electron Microscopy Sciences Supplier No. 775192
Lab mats with nanoscale silver particles bind with proteins to neutralize 99.9% of common microbes. Ensure consistant sample reproduction and results by significantly minimizing workspace contamination. Silver is used extensively in medical applications where microbial contamination is a concern. Mats are less than 1 mm thick and are backed with an anti-skid coating making them perfect for the benchtop or culture hood.
Size: 15" x 10"