Learn More
Medchemexpress LLC Silver bis(trifluoromethanesulfonyl)imide | 189114-61-2 | MFCD08457644 | 99.9% | 388.01 g·mol⁻¹ | C2AgF6NO4S2 | 1 G
SDP
Catalog No. 5000300116
Encompass_Preferred
Silver bis(trifluoromethanesulfonyl)imide is a silver(I) salt with the weakly coordinating bis(trifluoromethanesulfonyl)imide (TFSI) anion. It is intended for research use in applications that require a noncoordinating, fluorinated anion and high chemical purity, such as electrolyte development, ionic liquid studies, and silver-mediated syntheses.
Catalog No. 50-003-00116 Supplier Medchemexpress LLC Supplier No. HYW0210051G
Silver bis(trifluoromethanesulfonyl)imide is a silver(I) salt with the weakly coordinating bis(trifluoromethanesulfonyl)imide (TFSI) anion. It is intended for research use in applications that require a noncoordinating, fluorinated anion and high chemical purity, such as electrolyte development, ionic liquid studies, and silver-mediated syntheses.