Learn More
Medchemexpress LLC Silver bis(trifluoromethanesulfonyl)imide | 189114-61-2 | MFCD08457644 | 99.9% | 388.01 g·mol⁻¹ | C2AgF6NO4S2 | 10 G
SDP
Catalog No. 5000300941
Encompass_Preferred
Silver bis(trifluoromethanesulfonyl)imide is a silver salt used as a reagent and catalyst in organic synthesis and as a source of Ag+ for preparation of ionic liquids and other silver-based compounds. It is supplied as a high-purity solid for research and development chemistry applications.
  • High purity: 99.90% as supplied.
  • White to gray solid, easy to handle in laboratory settings.
  • Molecular weight 388.01 g·mol⁻¹; formula C2AgF6NO4S2.
  • Functions as a Lewis acid catalyst and source of Ag+.
  • Suitable for synthesis of ionic liquids and silver salts.
Catalog No. 50-003-00941 Supplier Medchemexpress LLC Supplier No. HYW02100510G
Silver bis(trifluoromethanesulfonyl)imide is a silver salt used as a reagent and catalyst in organic synthesis and as a source of Ag+ for preparation of ionic liquids and other silver-based compounds. It is supplied as a high-purity solid for research and development chemistry applications.
  • High purity: 99.90% as supplied.
  • White to gray solid, easy to handle in laboratory settings.
  • Molecular weight 388.01 g·mol⁻¹; formula C2AgF6NO4S2.
  • Functions as a Lewis acid catalyst and source of Ag+.
  • Suitable for synthesis of ionic liquids and silver salts.