Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Silver bis(trifluoromethanesulfonyl)imide | 189114-61-2 | MFCD08457644 | 99.9% | 388.01 g·mol⁻¹ | C2AgF6NO4S2 | 250 MG
SDP

Catalog No. 5000302592
Encompass_Preferred

Silver bis(trifluoromethanesulfonyl)imide is a silver salt used as a catalyst in organic synthesis and as a p-type dopant in perovskite solar cells. It enables C-3 regioselective alkylation of indole derivatives via conjugate addition and is supplied at high reported purity for research applications.

  • High purity: 99.9% (as supplied).
  • Effective catalyst for organic synthesis reactions.
  • Promotes C-3 regioselective alkylation of indoles.
  • Useful as a solid-state dye sensitizer and p-type dopant for perovskites.
  • Available in small pack sizes suitable for laboratory research.
  • Molecular weight 388.01 g·mol⁻¹ and formula C2AgF6NO4S2.

Catalog No. 50-003-02592 Supplier Medchemexpress LLC Supplier No. 250MGHYW021005
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Silver bis(trifluoromethanesulfonyl)imide is a silver salt used as a catalyst in organic synthesis and as a p-type dopant in perovskite solar cells. It enables C-3 regioselective alkylation of indole derivatives via conjugate addition and is supplied at high reported purity for research applications.

  • High purity: 99.9% (as supplied).
  • Effective catalyst for organic synthesis reactions.
  • Promotes C-3 regioselective alkylation of indoles.
  • Useful as a solid-state dye sensitizer and p-type dopant for perovskites.
  • Available in small pack sizes suitable for laboratory research.
  • Molecular weight 388.01 g·mol⁻¹ and formula C2AgF6NO4S2.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.