Learn More
Medchemexpress LLC Silver bis(trifluoromethanesulfonyl)imide | 189114-61-2 | MFCD08457644 | 99.9% | 388.01 g/mol | C2AgF6NO4S2 | 5 G
SDP
Catalog No. 5000300919
Encompass_Preferred
Silver bis(trifluoromethanesulfonyl)imide is a silver-based salt used as a catalyst and reagent in organic synthesis. Supplied as a solid for research use, it is typically handled under inert or anhydrous conditions and is used to promote electrophilic activation and to form ionic species in specialized reactions.
  • High purity for reproducible reactions.
  • Solid, easy-to-handle form suitable for laboratory use.
  • Effective catalyst for a variety of organic transformations.
  • Useful for generating ionic species and promoting electrophilic activation.
  • Provided in small quantities to support research-scale experiments.
Catalog No. 50-003-00919 Supplier Medchemexpress LLC Supplier No. HYW0210055G
Silver bis(trifluoromethanesulfonyl)imide is a silver-based salt used as a catalyst and reagent in organic synthesis. Supplied as a solid for research use, it is typically handled under inert or anhydrous conditions and is used to promote electrophilic activation and to form ionic species in specialized reactions.
  • High purity for reproducible reactions.
  • Solid, easy-to-handle form suitable for laboratory use.
  • Effective catalyst for a variety of organic transformations.
  • Useful for generating ionic species and promoting electrophilic activation.
  • Provided in small quantities to support research-scale experiments.