Learn More
Perkin Elmer US LLC Silver Capsule - Qty. 96
Catalog No. 509052191
Encompass_Preferred
These silver capsules also come in a light weight version that produce 40% less ash than standard weight capsules and provide a corresponding reduction in blank values.
Catalog No. 50-905-2191 Supplier Perkin Elmer US LLC Supplier No. N2411364
These silver capsules also come in a light weight version that produce 40% less ash than standard weight capsules and provide a corresponding reduction in blank values.