Learn More
Strem, An Ascensus Company CAS# 534-16-7. 10g. Silver carbonate (99+%-Ag). MFCD00003403
SDP
Supplier:  Strem, An Ascensus Company 93470610G
Encompass
CAS# 534-16-7. 10g. Silver carbonate (99+%-Ag). MFCD00003403. Molecular Weight: 275.75. Molecular Formula: Ag2CO3. Color/form: yellow to pale green pwdr. Strem# 93-4706. http://www.strem.com/catalog/v/93-4706/
Catalog No. 50-138-5561