Learn More
Medchemexpress LLC Silver chloride | 7783-90-6 | MFCD00003399 | 99.5% | 143.32 g/mol | AgCl | 10 G
SDP
Catalog No. 5000301516
Encompass_Preferred
Silver chloride (AgCl) is an inorganic reagent used in analytical and laboratory research. This high-purity (99.5%) solid is commonly used as a reference material, in electrochemistry, in silver-based syntheses, and in imaging applications. It is supplied as a fine powder in multiple package sizes for research and analytical use.
  • High purity (99.5%).
  • Low solubility in water, stable under normal laboratory conditions.
  • Suitable for analytical, electrochemical, and synthetic applications.
  • Available in multiple package sizes for flexible use.
  • Provided with safety and handling documentation for laboratory use.
Catalog No. 50-003-01516 Supplier Medchemexpress LLC Supplier No. HYW01992910G
Silver chloride (AgCl) is an inorganic reagent used in analytical and laboratory research. This high-purity (99.5%) solid is commonly used as a reference material, in electrochemistry, in silver-based syntheses, and in imaging applications. It is supplied as a fine powder in multiple package sizes for research and analytical use.
  • High purity (99.5%).
  • Low solubility in water, stable under normal laboratory conditions.
  • Suitable for analytical, electrochemical, and synthetic applications.
  • Available in multiple package sizes for flexible use.
  • Provided with safety and handling documentation for laboratory use.