Learn More
Medchemexpress LLC Silver chloride | 7783-90-6 | MFCD00003399 | 99.5% | 143.32 g/mol | AgCl | 50 G
SDP
Catalog No. 5000301518
Encompass_Preferred
Silver chloride (AgCl) is an inorganic reagent commonly used in analytical and laboratory research. This white crystalline solid is sparingly soluble in water and is employed in electrode preparation, photographic processes, inorganic synthesis, and analytical assays that require a high-purity silver salt.
  • High purity (99.5%) for research applications.
  • Well-defined molecular weight of 143.32 g/mol.
  • Low solubility useful for precipitation and analytical separations.
  • Suitable for electrode and photographic material preparation.
  • Supplied as a stable, easy-to-handle white crystalline solid.
Catalog No. 50-003-01518 Supplier Medchemexpress LLC Supplier No. HYW01992950G
Silver chloride (AgCl) is an inorganic reagent commonly used in analytical and laboratory research. This white crystalline solid is sparingly soluble in water and is employed in electrode preparation, photographic processes, inorganic synthesis, and analytical assays that require a high-purity silver salt.
  • High purity (99.5%) for research applications.
  • Well-defined molecular weight of 143.32 g/mol.
  • Low solubility useful for precipitation and analytical separations.
  • Suitable for electrode and photographic material preparation.
  • Supplied as a stable, easy-to-handle white crystalline solid.