Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Silver chloride, 95%

Silver plating

Manufacturer:  Alfa Aesar™ 8734214

Quantity 25g

 View more versions of this product

Catalog No. AA8734214


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7783-90-6
143.32
UN3077
Silver chloride
AgCl
MFCD00003399
14,8509

Specifications

7783-90-6
AgCl
MFCD00003399
14,8509
5.56g/mL
2.071
1550°C
455°C
Silver chloride
Coarse Powder
143.32
UN3077
Practically insoluble in water; Soluble in ammonia, solutions of alkali cyanides, thiosulfates, ammonium carbonates; Insoluble in alcohol, dilute acids
Odorless
Light Sensitive
White
25g
95%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H290
May be corrosive to metals.

GHS P Statement
P234-P390-P406
Keep only in original container.
Absorb spillage to prevent material damage.
Store in corrosive resistant container with a resistant inner liner.

Warning

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

RTECSNumber : VW3563000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title