Learn More
Strem, An Ascensus Company CAS# 7783-90-6. 50g. Silver chloride (99.9%-Ag). MFCD00003399
SDP
Catalog No. 5090115393
Encompass
CAS# 7783-90-6. 50g. Silver chloride (99.9%-Ag). MFCD00003399. Molecular Weight: 143.32. Molecular Formula: AgCl. Color/form: white pwdr. Strem# 93-4707. http://www.strem.com/catalog/v/93-4707/
Catalog No. 50-901-15393 Supplier Strem, An Ascensus Company Supplier No. 93470750G
CAS# 7783-90-6. 50g. Silver chloride (99.9%-Ag). MFCD00003399. Molecular Weight: 143.32. Molecular Formula: AgCl. Color/form: white pwdr. Strem# 93-4707. http://www.strem.com/catalog/v/93-4707/