Learn More
Silver Chromate 99%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Color | Black to Gray |
| Physical Form | Powder |
| Quantity | 10 g |
| Linear Formula | Ag2CrO4 |
| Fieser | 01,1005 |
| Merck Index | 15, 8649 |
| SMILES | [O-][Cr](=O)(=O)[O-].[Ag+].[Ag+] |
| IUPAC Name | disilver;dioxido(dioxo)chromium |
| Formula Weight | 331.72 |
| Percent Purity | ≥98.5% (AG2CRO4) |
| Show More |
Safety and Handling
| Hazard Category | HYGROSCOPIC |
| Hazard Statement | LIGHT SENSITIVE |
| EINECSNumber | 232-043-8 |
| TSCA | TSCA |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.