Learn More
Electron Microscopy Sciences EMS Silver Conductive 18DB70X Coating 15 GR
SDP
Catalog No. 50980328
Encompass
Silver Conductive Coating 18DB70X is a direct substitute for Silver Conductive Fluid 416, which is no longer available. This material was specially formulated for use in geographic areas that demand the use of low VOC (Volatile Organic Compounds) products. It also provides low ohms at very thin dry film thickness on almost any surface, and exhibits excellent long term shielding and grounding properties.
Catalog No. 50-980-328 Supplier Electron Microscopy Sciences Supplier No. 1268415
Silver Conductive Coating 18DB70X is a direct substitute for Silver Conductive Fluid 416, which is no longer available. This material was specially formulated for use in geographic areas that demand the use of low VOC (Volatile Organic Compounds) products. It also provides low ohms at very thin dry film thickness on almost any surface, and exhibits excellent long term shielding and grounding properties.