Learn More
Ted Pella Inc Silver Conductive Epoxy
SDP
Catalog No. NC9959045
Encompass
Strong, highly conductive epoxy for solderless connections and repairs of circuit boards, static discharge shielding, grounding and conductive paths. Properties: Bonding surface mount components, High strength conductive bonds, Quick, low or no heat cure – air dries, Rework or remove with hot soldering iron, Excellent electrical conductivity, and 2-part epoxy, reusable applicator with stir stick, 7g each.
Catalog No. NC9959045 Supplier Ted Pella Inc Supplier No. 16043
Strong, highly conductive epoxy for solderless connections and repairs of circuit boards, static discharge shielding, grounding and conductive paths. Properties: Bonding surface mount components, High strength conductive bonds, Quick, low or no heat cure – air dries, Rework or remove with hot soldering iron, Excellent electrical conductivity, and 2-part epoxy, reusable applicator with stir stick, 7g each.