Learn More
Structure Probe Inc SILVER CONDUCTIVE PAINT 1 OZ
SDP
Catalog No. NC9844600
Encompass
SPI Conductive Silver Paint,1 Troy oz.(31.1g),Use : for affixing samples to specimen mounts,or for attaching specimens in the microprobe and tilting stages of transmission electron microscopes (TEMs),High percent of silver solids
Catalog No. NC9844600 Supplier Structure Probe Inc Supplier No. 05002AB
SPI Conductive Silver Paint,1 Troy oz.(31.1g),Use : for affixing samples to specimen mounts,or for attaching specimens in the microprobe and tilting stages of transmission electron microscopes (TEMs),High percent of silver solids