Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver Cyanide, Powder, Purified, 99%, Spectrum™ Chemical
SDP

Catalog No. 18613130 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
100 g
125 g
500 g
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-613-130 500 g Amber Glass Bottle
18-613-127 25 g Amber Glass Bottle
18-613-128 100 g Amber Glass Bottle
18-613-129 125 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-613-130 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S1060500GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 506-64-9

Specifications

Melting Point 320°C
Color Grayish White
CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 500 g
Molecular Formula AgCN
Linear Formula AgCN
Synonym Argentous cyanide, Kyanid stribrny, Silver cyanide (Ag(CN)), Silver(1+) cyanide
Solubility Information Soluble in Water
IUPAC Name Silver(1+) cyanide
Molecular Weight (g/mol) 133.9
PubChem CID 10475
Formula Weight 133.90 g/mol
Percent Purity 99%
Grade Purified
Odor Odorless
Packaging Amber Glass Bottle
Appearance Grayish White
Chemical Name or Material Silver Cyanide, Powder
Show More Show Less
Recommended Storage Light sensitive, Keep tightly closed in light-resistant containers

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.