Learn More
Medchemexpress LLC Silver diethyldithiocarbamate | 1470-61-7 | MFCD00004929 | 256.1 g/mol | C5H10AgNS2 | 50 G
SDP
Catalog No. 5000300176
Encompass_Preferred
Silver diethyldithiocarbamate is a silver(I) salt of diethyldithiocarbamate used primarily as a reagent in analytical chemistry and as a metal-complexing agent. It is typically a white to pale-yellow solid with molecular formula C5H10AgNS2 and molecular weight 256.1 g/mol (CAS 1470-61-7).
  • Used for colorimetric determination of arsenic in analytical methods.
  • Forms stable complexes with transition metal ions.
  • Available as a solid powder suitable for reagent preparation and analysis.
  • Soluble in select organic solvents such as pyridine.
  • High purity grades (commonly reported as ≥98% by suppliers) suitable for analytical use.
Catalog No. 50-003-00176 Supplier Medchemexpress LLC Supplier No. HYW03467450G
Silver diethyldithiocarbamate is a silver(I) salt of diethyldithiocarbamate used primarily as a reagent in analytical chemistry and as a metal-complexing agent. It is typically a white to pale-yellow solid with molecular formula C5H10AgNS2 and molecular weight 256.1 g/mol (CAS 1470-61-7).
  • Used for colorimetric determination of arsenic in analytical methods.
  • Forms stable complexes with transition metal ions.
  • Available as a solid powder suitable for reagent preparation and analysis.
  • Soluble in select organic solvents such as pyridine.
  • High purity grades (commonly reported as ≥98% by suppliers) suitable for analytical use.