Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Silver diethyldithiocarbamate | 1470-61-7 | MFCD00004929 | 99.2% | 256.14 g·mol^-1 | C5H10AgNS2 | 100 G
SDP

Catalog No. 5000299530
Encompass_Preferred

Silver diethyldithiocarbamate is a silver(I) complex reagent used in analytical chemistry and life-science research. It forms stable complexes with metal ions and is commonly applied in colorimetric determinations and the detection of metals such as arsenic, copper, and iron. Provided as a high-purity solid for laboratory use.

  • High purity (99.2%).
  • Molecular formula C5H10AgNS2; molecular weight 256.14 g·mol^-1.
  • Suitable for colorimetric and metal-detection assays.
  • Chelates and forms stable metal complexes.
  • Solid reagent; protect from light and store under nitrogen at 4°C.
  • Available in multiple pack sizes for laboratory use.

Catalog No. 50-002-99530 Supplier Medchemexpress LLC Supplier No. HYW034674100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Silver diethyldithiocarbamate is a silver(I) complex reagent used in analytical chemistry and life-science research. It forms stable complexes with metal ions and is commonly applied in colorimetric determinations and the detection of metals such as arsenic, copper, and iron. Provided as a high-purity solid for laboratory use.

  • High purity (99.2%).
  • Molecular formula C5H10AgNS2; molecular weight 256.14 g·mol^-1.
  • Suitable for colorimetric and metal-detection assays.
  • Chelates and forms stable metal complexes.
  • Solid reagent; protect from light and store under nitrogen at 4°C.
  • Available in multiple pack sizes for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.