Learn More
Medchemexpress LLC Silver diethyldithiocarbamate | 1470-61-7 | MFCD00004929 | 99.2% | 256.14 g/mol | C5H10AgNS2 | 10 G
SDP
Catalog No. 5000299534
Encompass_Preferred
Silver diethyldithiocarbamate is the silver salt of diethyldithiocarbamic acid supplied as a high-purity powder for laboratory analytical use. It acts as a chelating reagent that forms colored complexes with metal ions and is used in colorimetric and spectroscopic metal determinations.
  • High purity suitable for analytical applications.
  • Forms stable complexes with transition metals for colorimetric assays.
  • Available in multiple pack sizes to support varied laboratory needs.
  • Documentation available (COA and SDS) for quality and safety verification.
  • Solid powder form for convenient reagent preparation and storage.
Catalog No. 50-002-99534 Supplier Medchemexpress LLC Supplier No. HYW03467410G
Silver diethyldithiocarbamate is the silver salt of diethyldithiocarbamic acid supplied as a high-purity powder for laboratory analytical use. It acts as a chelating reagent that forms colored complexes with metal ions and is used in colorimetric and spectroscopic metal determinations.
  • High purity suitable for analytical applications.
  • Forms stable complexes with transition metals for colorimetric assays.
  • Available in multiple pack sizes to support varied laboratory needs.
  • Documentation available (COA and SDS) for quality and safety verification.
  • Solid powder form for convenient reagent preparation and storage.