Learn More
Medchemexpress LLC Silver diethyldithiocarbamate | 1470-61-7 | MFCD00004929 | 99.2% | 256.14 g·mol⁻¹ | C5H10AgNS2 | 25 G
SDP
Catalog No. 5000303041
Encompass_Preferred
Silver diethyldithiocarbamate is a silver salt of the diethyldithiocarbamate ligand used as a chelating reagent in analytical chemistry to form stable complexes with metal ions for spectroscopic analysis. It is supplied as a solid with high stated purity and is suitable for laboratory use in metal detection and related assays.
  • Used as a reagent and chelating agent for metal ion detection.
  • High purity suitable for UV-Vis spectroscopy and analytical work.
  • Light yellow to light brown solid appearance.
  • Molecular weight 256.14 g·mol⁻¹; formula C5H10AgNS2.
  • Provided in 25 g packaging for laboratory-scale use.
Catalog No. 50-003-03041 Supplier Medchemexpress LLC Supplier No. 25GHYW034674
Silver diethyldithiocarbamate is a silver salt of the diethyldithiocarbamate ligand used as a chelating reagent in analytical chemistry to form stable complexes with metal ions for spectroscopic analysis. It is supplied as a solid with high stated purity and is suitable for laboratory use in metal detection and related assays.
  • Used as a reagent and chelating agent for metal ion detection.
  • High purity suitable for UV-Vis spectroscopy and analytical work.
  • Light yellow to light brown solid appearance.
  • Molecular weight 256.14 g·mol⁻¹; formula C5H10AgNS2.
  • Provided in 25 g packaging for laboratory-scale use.