Learn More
Hach Company Silver Diethyldithiocarbamate, 25 g
Catalog No. 501995041 Shop All Hach Company Products
Encompass
ACS Grade, CAS No. 1470-61-7 (C2H5)2NCS2Ag, FW = 256.13. Suitability for determination of arsenic: Passes test
Catalog No. 50-199-5041 Supplier Hach Company Supplier No. 1447624
ACS Grade, CAS No. 1470-61-7 (C2H5)2NCS2Ag, FW = 256.13. Suitability for determination of arsenic: Passes test