Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Silver, dispersion

Catalog No. 73646525G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
73-646-525G 25 g
73-646-5100G -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 73-646-525G Supplier Sigma Aldrich Supplier No. 73646525G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Silver is a good conductor and is resistant to oxidation. It is used in printed conductors and printable inks (based on silver nanoparticles).Other large scale applications of nanosilver include metallization of solar cells, thin film electronics, sensors, and catalysts. Curing Temperature: 100-150 CRecommended substrates: Plastic filmsRecommended washing solvent: Polar Solvents: Ethyl Alcohol, IPA etc.Product of ANP, Inc.

Specifications

Density 1.45 g/mL ±0.05 g/mL (at 25°C)
Quantity 25 g
Molecular Formula Ag
Linear Formula Ag
MDL Number MFCD00003397
Synonym Silverjet DGP-40 LT-15 C; silver ink
Molecular Weight (g/mol) 107.87
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.