Learn More
Seracare Life Sciences Inc SILVER ENHANC KIT-MEMBRANES
Catalog No. 5067404
Encompass_Preferred
KPL's Silver Enhancer Kits are available for use in membrane based applications. This simple yet sensitive system is optimized for use with KPL's gold conjugates for the amplification of gold particles. Silver builds up on the surface of the gold particles as a result of the reduction of silver from one solution to another. Silver Enhancer kits offer low background, sensitivity to the picogram level and slow, controlled development. 15-25 minutes development for microscopy applications.
This item is not returnable. View return policy
Catalog No. 50-674-04 Supplier Seracare Life Sciences Inc Supplier No. 54500012
KPL's Silver Enhancer Kits are available for use in membrane based applications. This simple yet sensitive system is optimized for use with KPL's gold conjugates for the amplification of gold particles. Silver builds up on the surface of the gold particles as a result of the reduction of silver from one solution to another. Silver Enhancer kits offer low background, sensitivity to the picogram level and slow, controlled development. 15-25 minutes development for microscopy applications.