Learn More
Strem, An Ascensus Company CAS# 7440-22-4. 5g. Silver flake (99.9%). MFCD00003397
SDP
Supplier:  Strem, An Ascensus Company 9347605G
Encompass
CAS# 7440-22-4. 5g. Silver flake (99.9%). MFCD00003397. Molecular Weight: 107.87. Molecular Formula: Ag. Color/form: 100% < 20 micron Strem# 93-4760. http://www.strem.com/catalog/v/93-4760/
Catalog No. 50-901-15444