Learn More
Sigma Aldrich Fine Chemicals Biosciences Silver foil, 25mm disks, thickness 0.15mm, as rolled, 99.95+% | 7440-22-4 | MFCD00003397 | 10EA
Catalog No. 501853193
Encompass
Silver foil, 25mm disks, thickness 0.15mm, as rolled, 99.95+% | Purity: 99.95+ % | Mol Wt: 107.87 | 7440-22-4 | MFCD00003397 | 10EA
Catalog No. 50-185-3193 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GF8115008310EA
Silver foil, 25mm disks, thickness 0.15mm, as rolled, 99.95+% | Purity: 99.95+ % | Mol Wt: 107.87 | 7440-22-4 | MFCD00003397 | 10EA