Learn More
Sigma Aldrich Fine Chemicals Biosciences Silver foil, 6mm disks, thickness 0.1mm, annealed, 99.95+% | 7440-22-4 | MFCD00003397 | 10EA
Catalog No. 501853073
Encompass
Silver foil, 6mm disks, thickness 0.1mm, annealed, 99.95+% | Purity: 99.95+ % | Mol Wt: 107.87 | 7440-22-4 | MFCD00003397 | 10EA
Catalog No. 50-185-3073 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GF7547624510EA
Silver foil, 6mm disks, thickness 0.1mm, annealed, 99.95+% | Purity: 99.95+ % | Mol Wt: 107.87 | 7440-22-4 | MFCD00003397 | 10EA