Learn More
Sigma Aldrich Fine Chemicals Biosciences Silver foil, light tested, 25x25mm, thickness 0.075mm, as rolled, 99.95+% | 7440-22-4 | MFCD00003397 | 5EA
Catalog No. 501853397
Encompass
Silver foil, light tested, 25x25mm, thickness 0.075mm, as rolled, 99.95+% | Purity: >=99.95% | Mol Wt: 107.87 | 7440-22-4 | MFCD00003397 | 5EA
Catalog No. 50-185-3397 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GF927932685EA
Silver foil, light tested, 25x25mm, thickness 0.075mm, as rolled, 99.95+% | Purity: >=99.95% | Mol Wt: 107.87 | 7440-22-4 | MFCD00003397 | 5EA