Learn More
Sigma Aldrich Fine Chemicals Biosciences Silver foil, thickness 0.015 mm, size 100 x 100 mm, purity 99.95+% | 7440-22-4 | MFCD00003397 | 5EA
Catalog No. 501853095
Encompass
Silver foil, thickness 0.015 mm, size 100 x 100 mm, purity 99.95+% | Purity: >=99.95% | Mol Wt: 107.87 | 7440-22-4 | MFCD00003397 | 5EA
Catalog No. 50-185-3095 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GF763508545EA
Silver foil, thickness 0.015 mm, size 100 x 100 mm, purity 99.95+% | Purity: >=99.95% | Mol Wt: 107.87 | 7440-22-4 | MFCD00003397 | 5EA